Assay Details
Target Gene Details
Entrez Gene ID: | 327814 |
Gene Name: | protein tyrosine phosphatase, receptor type, f polypeptide (PTPRF), interacting protein (liprin), alpha 2 |
Gene Aliases: |
5330438O12, B230207K17Rik, E130120L08Rik, mKIAA4112 |
Location: |
Chr.10:106468872-106933468 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 21 - Intron 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ppfia2 | NM_001205341.1 | NP_001192270.1 | ||
NM_177373.4 | NP_796347.2 | |||
AK220491.1 | BAD90289.1 | |||
BC065133.1 | AAH65133.1 | |||
EU568870.1 |