Assay Details
Target Gene Details
Entrez Gene ID: | 216443 |
Gene Name: | methionine-tRNA synthetase |
Gene Aliases: |
Metrs, Mtrns |
Location: |
Chr.10:127296221-127311786 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mars | NM_001003913.2 | NP_001003913.1 | ||
NM_001171582.1 | NP_001165053.1 | |||
AK181712.1 | ||||
BC079643.1 | AAH79643.1 |
More Information
Set Membership: |
Intragenic Non-exonic |