Assay Details
Target Gene Details
Entrez Gene ID: | 68652 |
Gene Name: | TGF-beta activated kinase 1/MAP3K7 binding protein 2 |
Gene Aliases: |
1110030N06Rik, A530078N03Rik, Map3k7ip2, mKIAA0733 |
Location: |
Chr.10:7905648-7956225 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tab2 | NM_138667.3 | NP_619608.1 | ||
AB093262.1 | BAC41446.1 | |||
AK217747.1 | ||||
AY093701.1 | AAM10487.1 | |||
BC004072.1 | AAH04072.1 | |||
BC004813.1 | AAH04813.1 |