Assay Details
Target Gene Details
Entrez Gene ID: | 216136 |
Gene Name: | ilvB (bacterial acetolactate synthase)-like |
Gene Aliases: |
5830463I21, AI415009 |
Location: |
Chr.10:78574345-78584502 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 13 - Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ilvbl | NM_173751.4 | NP_776112.1 | ||
AK030933.1 | 9 | 987 | ||
AK087956.1 | BAC40057.1 | |||
AK146075.1 | BAE26879.1 | |||
AK169899.1 | ||||
AK184668.1 | ||||
AK218076.1 | 1 | 343 | ||
BC056459.1 | AAH56459.1 | |||
CK789563.1 |
More Information
Set Membership: |
Intragenic Non-exonic |