Assay Details
Target Gene Details
Entrez Gene ID: | 52665 |
Gene Name: | enoyl Coenzyme A hydratase domain containing 1 |
Gene Aliases: |
1700028A24Rik, AI314462, AI930038, D10Ertd667e, MMCD |
Location: |
Chr.10:29311507-29346661 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Echdc1 | NM_001110195.1 | 2 | 283 | NP_001103665.1 |
NM_025855.4 | 2 | 385 | NP_080131.4 | |
AK006444.1 | 2 | 262 | BAB24592.2 | |
AK016260.1 | 2 | 327 | ||
AK028775.1 | 2 | 282 | BAC26112.1 | |
AK150932.1 | 2 | 267 | BAE29969.1 | |
AK152285.1 | 2 | 267 | BAE31098.1 | |
AK153454.1 | 2 | 267 | BAE32007.1 | |
AK166589.1 | 2 | 307 | BAE38876.1 | |
AK166634.1 | 2 | 307 | BAE38907.1 | |
AK166660.1 | 2 | 307 | BAE38924.1 | |
BC066183.1 | 2 | 250 | AAH66183.1 | |
BY325699.1 | ||||
CJ051963.1 | 2 | 386 |
More Information
Set Membership: |
Intragenic Exonic |