Assay Details
Target Gene Details
Entrez Gene ID: | 52665 |
Gene Name: | enoyl Coenzyme A hydratase domain containing 1 |
Gene Aliases: |
1700028A24Rik, AI314462, AI930038, D10Ertd667e, MMCD |
Location: |
Chr.10:29311507-29346661 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Echdc1 | NM_001110195.1 | NP_001103665.1 | ||
AK150932.1 | BAE29969.1 | |||
AK152285.1 | BAE31098.1 | |||
AK153454.1 | BAE32007.1 | |||
BY325699.1 |
More Information
Set Membership: |
Intragenic Non-exonic |