Assay Details
Target Gene Details
Entrez Gene ID: | 59012 |
Gene Name: | monooxygenase, DBH-like 1 |
Gene Aliases: |
3230402N08Rik, MNCb-5203 |
Location: |
Chr.10:24223517-24302783 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Moxd1 | NM_021509.5 | NP_067484.2 | ||
AB041606.1 | BAA95089.1 | |||
AK014340.1 | BAB29283.1 | |||
AK030274.1 | BAC26871.1 | |||
AK081586.1 | BAC38265.1 | |||
AK162030.1 | ||||
BC025892.1 | AAH25892.1 | |||
BC057652.1 | AAH57652.1 |
More Information
Set Membership: |
Intragenic Non-exonic |