Assay Details
Target Gene Details
Entrez Gene ID: | 237339 |
Gene Name: | l(3)mbt-like 3 (Drosophila) |
Gene Aliases: |
AI481284, MBT-1 |
Location: |
Chr.10:26274470-26375989 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
L3mbtl3 | NM_172787.2 | 18 | 1908 | NP_766375.1 |
AK030412.1 | 5 | 506 | BAC26951.1 | |
AK045667.1 | 18 | 1909 | BAC32449.1 | |
AK049597.1 | 18 | 1943 | ||
AK129443.1 | 14 | 1453 | BAC98253.1 | |
BC082309.1 | 17 | 1778 | AAH82309.1 | |
BC085192.1 | 17 | 2013 |
More Information
Set Membership: |
Intragenic Exonic |