Assay Details
Target Gene Details
Entrez Gene ID: | 237313 |
Gene Name: | interleukin 20 receptor, alpha |
Gene Aliases: |
E230031K19 |
Location: |
Chr.10:19711673-19760053 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Il20ra | NM_172786.2 | 3 | 405 | NP_766374.1 |
AK054215.1 | 3 | 406 | BAC35695.1 | |
BC056628.1 | 3 | 389 | AAH56628.1 | |
EF592560.1 | 4 | 483 |
More Information
Set Membership: |
Intragenic Exonic |