Assay Details
Target Gene Details
Entrez Gene ID: | 237313 |
Gene Name: | interleukin 20 receptor, alpha |
Gene Aliases: |
E230031K19 |
Location: |
Chr.10:19711673-19760053 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Il20ra | NM_172786.2 | NP_766374.1 | ||
AK054215.1 | BAC35695.1 | |||
BC056628.1 | AAH56628.1 | |||
EF592560.1 |
More Information
Set Membership: |
Intragenic Non-exonic |