Assay Details
Target Gene Details
Entrez Gene ID: | 15214 |
Gene Name: | hairy/enhancer-of-split related with YRPW motif 2 |
Gene Aliases: |
CHF1, Herp1, Hrt2, bHLHb32, hesr2 |
Location: |
Chr.10:30832359-30842783 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hey2 | NM_013904.1 | NP_038932.1 | ||
AB093589.1 | BAC55066.1 | |||
AF172287.1 | AAF14546.1 | |||
AF173902.1 | AAF20174.1 | |||
AF232240.1 | AAF37298.1 | |||
AJ271867.1 | CAB71346.1 | |||
AK031506.1 | BAC27428.1 | |||
AK158000.1 | BAE34310.1 | |||
BC103575.1 | AAI03576.1 | |||
BC103576.1 | AAI03577.1 | |||
BC105651.1 | ||||
BC105652.1 |
More Information
Set Membership: |
Intragenic Non-exonic |