Assay Details
Target Gene Details
Entrez Gene ID: | 74053 |
Gene Name: | glutamate receptor interacting protein 1 |
Gene Aliases: |
4931400F03Rik, GRIP, eb |
Location: |
Chr.10:119453835-120087267 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Grip1 | NM_001277292.1 | NP_001264221.1 | ||
NM_001277293.1 | NP_001264222.1 | |||
NM_133442.2 | NP_597699.1 | |||
AB051562.1 | BAB46931.1 | |||
AK220553.1 | BAD90538.1 | |||
BC072632.1 | AAH72632.1 |