Assay Details
Target Gene Details
Entrez Gene ID: | 237558 |
Gene Name: | myelin regulatory factor-like |
Gene Aliases: |
Gm239 |
Location: |
Chr.10:116776545-116896879 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Myrfl | NM_001033333.3 | 19 | 2384 | NP_001028505.1 |
AK144383.1 | 19 | 2385 | BAE25858.1 | |
AK144411.1 | 19 | 2385 | BAE25877.1 | |
BC150915.1 | 19 | 2188 | ||
BC150923.1 | 19 | 2188 |