Assay Details
Target Gene Details
Entrez Gene ID: | 18483 |
Gene Name: | paralemmin |
Gene Aliases: |
- |
Location: |
Chr.10:79793351-79820896 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Palm | NM_001161747.1 | NP_001155219.1 | ||
NM_023128.4 | NP_075617.3 | |||
AK009200.1 | ||||
AK037044.1 | BAC29683.1 | |||
AK082163.1 | BAC38429.1 | |||
AK129107.1 | BAC97917.1 | |||
AK133733.1 | 7 | 2263 | BAE21809.1 | |
AK162411.1 | BAE36902.1 | |||
BC015297.1 | AAH15297.1 | |||
Y14771.1 | CAB37403.1 |