Assay Details
Target Gene Details
Entrez Gene ID: | 17311 |
Gene Name: | kit ligand |
Gene Aliases: |
Clo, Con, Gb, Kitlg, Mgf, SCF, SF, SLF, Sl, blz, contrasted |
Location: |
Chr.10:100015809-100100416 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kitl | NM_013598.2 | NP_038626.1 | ||
AK018777.1 | BAB31402.1 | |||
AK134301.1 | BAE22091.1 | |||
AK165233.1 | BAE38091.1 | |||
BC011322.1 | AAH11322.1 | |||
M57647.1 | AAA39538.1 | |||
M64262.1 | AAA39378.1 | |||
S40364.1 | AAB22554.2 | |||
S40534.1 | AAB22555.2 | |||
S40536.1 | AAB22556.2 | |||
U44725.1 | AAC52447.1 | |||
X68989.1 | CAA48778.1 | |||
X95379.1 | CAA64666.1 | |||
X95380.1 | ||||
X95381.1 | CAA64667.1 | |||
X99322.1 | CAA67698.1 | |||
Y10287.1 | CAA71329.1 |