Assay Details
Target Gene Details
Entrez Gene ID: | 67074 |
Gene Name: | MON2 homolog (yeast) |
Gene Aliases: |
2610528O22Rik, AW495628, Sf21, mKIAA1040 |
Location: |
Chr.10:122992060-123076505 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mon2 | NM_001163024.1 | 23 | 3244 | NP_001156496.1 |
NM_001163025.1 | 22 | 3244 | NP_001156497.1 | |
NM_153395.2 | 22 | 3241 | NP_700444.2 | |
AB014059.1 | 22 | 2763 | BAC11706.1 | |
AK011826.1 | BAC25345.1 | |||
AK012182.2 | 2 | 181 | ||
AK122425.1 | 23 | 3244 | BAC65707.1 | |
AK191210.1 | 2 | 205 | ||
AK194501.1 | 2 | 165 | ||
BC150943.1 | 22 | 2765 | ||
CV563363.1 | 1 | 104 | ||
CX224591.1 | 7 | 635 |
More Information
Set Membership: |
Intragenic Exonic |