Assay Details
Target Gene Details
Entrez Gene ID: | 67074 |
Gene Name: | MON2 homolog (yeast) |
Gene Aliases: |
2610528O22Rik, AW495628, Sf21, mKIAA1040 |
Location: |
Chr.10:122992060-123076505 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mon2 | NM_001163024.1 | 20 | 2949 | NP_001156496.1 |
NM_001163025.1 | 20 | 2952 | NP_001156497.1 | |
NM_153395.2 | 20 | 2949 | NP_700444.2 | |
AB014059.1 | 20 | 2471 | BAC11706.1 | |
AK122425.1 | 20 | 2949 | BAC65707.1 | |
BC150943.1 | 20 | 2473 | ||
CX122556.1 | 7 | 563 | ||
CX224591.1 | 5 | 343 |
More Information
Set Membership: |
Intragenic Exonic |