Assay Details
Target Gene Details
Entrez Gene ID: | 380629 |
Gene Name: | headcase homolog (Drosophila) |
Gene Aliases: |
Gm869, HDC, HDCL |
Location: |
Chr.10:17899042-17948067 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Heca | NM_001033432.3 | 3 | 1684 | NP_001028604.1 |
AK132340.1 | 3 | 1685 | BAE21115.1 |