Assay Details
Target Gene Details
Entrez Gene ID: | 54427 |
Gene Name: | DNA (cytosine-5-)-methyltransferase 3-like |
Gene Aliases: |
D6Ertd14e, ecat7 |
Location: |
Chr.10:78042287-78063622 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dnmt3l | NM_001081695.2 | NP_001075164.1 | ||
NM_001284197.1 | NP_001271126.1 | |||
NM_019448.4 | NP_062321.1 | |||
AF220524.1 | AAF73868.1 | |||
AJ404467.1 | CAB94726.1 | |||
AK010434.1 | BAB26936.1 | |||
BC083147.1 | AAH83147.1 | |||
EF051624.1 |
More Information
Set Membership: |
Intragenic Non-exonic |