Assay Details
Target Gene Details
Entrez Gene ID: | 16993 |
Gene Name: | leukotriene A4 hydrolase |
Gene Aliases: |
- |
Location: |
Chr.10:93453396-93484898 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lta4h | NM_008517.2 | NP_032543.2 | ||
AK032828.1 | ||||
AK134931.1 | BAE22342.1 | |||
AK179027.1 | ||||
AK180384.1 | ||||
BC021417.1 | AAH21417.1 |