Assay Details
Target Gene Details
Entrez Gene ID: | 14806 |
Gene Name: | glutamate receptor, ionotropic, kainate 2 (beta 2) |
Gene Aliases: |
AW124492, GluK2, Glur-6, Glur6, Glurbeta2 |
Location: |
Chr.10:49094842-49789250 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Grik2 | NM_001111268.1 | 9 | 1562 | NP_001104738.1 |
NM_010349.2 | 9 | 1562 | NP_034479.2 | |
BC131639.1 | 8 | 1243 | ||
BC131640.1 | 8 | 1243 | ||
D10054.1 | 9 | 1475 | BAA00943.1 | |
X66117.1 | 8 | 1299 | CAA46907.1 |
More Information
Set Membership: |
Intragenic Exonic |