Assay Details
Target Gene Details
Entrez Gene ID: | 74782 |
Gene Name: | glycosyltransferase 8 domain containing 2 |
Gene Aliases: |
1110021D20Rik |
Location: |
Chr.10:82650428-82694113 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Glt8d2 | NM_029102.3 | 5 | 539 | NP_083378.2 |
AK003894.1 | 5 | 540 | BAB23061.1 | |
AK040842.1 | 5 | 507 | BAC30718.1 | |
AK136351.1 | 5 | 520 | ||
BC082561.1 | 5 | 468 | AAH82561.1 |
More Information
Set Membership: |
Intragenic Exonic |