Assay Details
Target Gene Details
Entrez Gene ID: | 67074 |
Gene Name: | MON2 homolog (yeast) |
Gene Aliases: |
2610528O22Rik, AW495628, Sf21, mKIAA1040 |
Location: |
Chr.10:122992060-123076505 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 29 - Exon 29 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mon2 | NM_001163024.1 | NP_001156496.1 | ||
NM_001163025.1 | NP_001156497.1 | |||
NM_153395.2 | NP_700444.2 | |||
AB014059.1 | BAC11706.1 | |||
AK011826.1 | BAC25345.1 | |||
AK012182.2 | ||||
AK122425.1 | BAC65707.1 | |||
AK162859.1 | BAE37087.1 | |||
BC150943.1 | ||||
CV561694.1 |
More Information
Set Membership: |
Intragenic Non-exonic |