Assay Details
Target Gene Details
Entrez Gene ID: | 94221 |
Gene Name: | golgi associated PDZ and coiled-coil motif containing |
Gene Aliases: |
2210402P09Rik, AI844555, CAL, FIG, GOPC1, PIST |
Location: |
Chr.10:52337024-52382124 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gopc | NM_001199272.1 | 8 | 1406 | NP_001186201.1 |
NM_053187.3 | 7 | 1382 | NP_444417.2 | |
AB052838.1 | 7 | 1162 | BAB69946.1 | |
AF287893.1 | 7 | 1298 | AAG00571.1 | |
AK030427.1 | 8 | 1002 | BAC26958.1 | |
AK030637.1 | 8 | 1215 | BAC27058.1 | |
AK032613.1 | 8 | 1212 | BAC27951.1 | |
BC051171.1 | 7 | 1333 | AAH51171.1 |
More Information
Set Membership: |
Intragenic Exonic |