Assay Details
Target Gene Details
Entrez Gene ID: | 216157 |
Gene Name: | transmembrane protein 259 |
Gene Aliases: |
AC087114.7gm2, ORF61, R32184_3, mFLJ00277 |
Location: |
Chr.10:79977120-79984330 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tmem259 | NM_001003949.3 | 4 | 833 | NP_001003949.2 |
AY096036.1 | 4 | 543 | AAM34492.1 | |
AY096037.1 | 4 | 543 | AAM34493.1 | |
BC005494.1 | 3 | 356 | AAH05494.1 | |
BC052787.1 | 3 | 375 | AAH52787.1 | |
BC100297.1 | AAI00298.1 |
More Information
Set Membership: |
Intragenic Exonic |