Assay Details
Target Gene Details
Entrez Gene ID: | 66414 |
Gene Name: | NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 12 |
Gene Aliases: |
2410011G03Rik, AW112974 |
Location: |
Chr.10:94199009-94220948 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ndufa12 | NM_025551.3 | NP_079827.2 | ||
AK003131.1 | BAB22591.2 | |||
AK003322.1 | BAB22713.2 | |||
AK008876.1 | BAB25945.2 | |||
AK010460.1 | BAB26955.2 | |||
AK032774.1 | BAC28016.1 | |||
AK134765.1 | BAE22273.1 | |||
AK153240.1 | BAE31832.1 | |||
AK167943.1 | BAE39946.1 | |||
BC086922.1 | AAH86922.1 |
More Information
Set Membership: |
Intragenic Non-exonic |