Assay Details
Target Gene Details
Entrez Gene ID: | 404634 |
Gene Name: | H2A histone family, member Y2 |
Gene Aliases: |
macroH2A2 |
Location: |
Chr.10:61738647-61784104 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
H2afy2 | NM_207000.2 | NP_996883.1 | ||
AF336305.1 | AAK52472.1 | |||
AK018355.1 | ||||
AK020744.1 | BAB32199.1 | |||
AK032636.1 | BAC27963.1 | |||
AK078464.1 | BAC37288.1 | |||
AK165020.1 | BAE38003.1 | |||
BC045140.1 | AAH45140.1 | |||
BC046794.1 | AAH46794.1 | |||
BC107279.1 | AAI07280.1 | |||
BC107280.1 | AAI07281.1 | |||
BC127144.1 |
More Information
Set Membership: |
Intragenic Non-exonic |