Assay Details
Target Gene Details
Entrez Gene ID: | 56371 |
Gene Name: | fizzy/cell division cycle 20 related 1 (Drosophila) |
Gene Aliases: |
AW108046, Cdh1, FZR, FZR2, Fyr, HCDH, HCDH1 |
Location: |
Chr.10:81366879-81378370 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fzr1 | NM_019757.1 | NP_062731.1 | ||
AF083809.1 | AAD52029.1 | |||
AK051993.1 | ||||
AK081476.1 | ||||
AK154822.1 | BAE32854.1 | |||
AK155370.1 | BAE33224.1 | |||
AK163951.1 | BAE37547.1 | |||
AK172679.1 | 8 | 1496 | ||
AK181261.1 | ||||
AK184844.1 | ||||
BC006616.1 | AAH06616.1 | |||
FJ786633.2 |
More Information
Set Membership: |
Intragenic Non-exonic |