Assay Details
Target Gene Details
Entrez Gene ID: | 18245 |
Gene Name: | ornithine decarboxylase antizyme 1 |
Gene Aliases: |
AZ-1, AZ1, Antizyme, ODC-Az, Oaz |
Location: |
Chr.10:80826656-80829290 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Oaz1 | NM_001301034.1 | NP_001287963.1 | ||
NM_008753.4 | NP_032779.2 | |||
AK003452.1 | ||||
AK010615.1 | ||||
AK019155.1 | ||||
AK087942.1 | ||||
AK146571.1 | ||||
AK147167.1 | ||||
AK151635.1 | ||||
AK159849.1 | ||||
AK159874.1 | ||||
AK160491.1 | ||||
AK160598.1 | ||||
AK169201.1 | ||||
AK201392.1 | ||||
BC094287.1 | AAH94287.1 | |||
BG296334.1 | ||||
U52822.1 | AAB96329.1 |
More Information
Set Membership: |
Intragenic Non-exonic |