Assay Details
Target Gene Details
Entrez Gene ID: | 67331 |
Gene Name: | ATPase, class I, type 8B, member 3 |
Gene Aliases: |
1700042F02Rik, 1700056N23Rik, ATPIK, SAPLT |
Location: |
Chr.10:80519584-80539150 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Atp8b3 | NM_026094.3 | NP_080370.2 | ||
AK006684.1 | BAB24703.1 | |||
AK018868.1 | BAB31468.1 | |||
AK018877.1 | BAB31471.1 | |||
AY364445.1 | AAR12913.1 | |||
BC049570.1 | AAH49570.1 | |||
BC118977.1 | ||||
BC118978.1 |
More Information
Set Membership: |
Intragenic Non-exonic |