Assay Details
Target Gene Details
Entrez Gene ID: | 74392 |
Gene Name: | sperm antigen with calponin homology and coiled-coil domains 1-like |
Gene Aliases: |
4930470P14Rik, 4932439K10Rik, 9530057A13Rik, AI317249, Cytsa, mKIAA0376 |
Location: |
Chr.10:75212073-75312400 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Specc1l | NM_001145826.1 | 5 | 434 | NP_001139298.1 |
NM_153406.3 | 2 | 205 | NP_700455.3 | |
AB093236.1 | 5 | 434 | BAC41420.1 | |
AK035500.1 | 2 | 207 | ||
AK052716.1 | 2 | 206 | BAC35113.1 | |
AK172502.1 | 2 | 183 | ||
AY884297.1 | 2 | 206 | AAX84188.1 | |
BC072595.1 | 1 | 44 | AAH72595.1 |