Assay Details
Target Gene Details
Entrez Gene ID: | 67500 |
Gene Name: | cell division cycle and apoptosis regulator 1 |
Gene Aliases: |
2610511G16Rik, 9430036H15Rik, Carp1 |
Location: |
Chr.10:62743928-62792368 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccar1 | NM_026201.3 | 9 | 1087 | NP_080477.1 |
AF465615.1 | 9 | 978 | AAO17318.1 | |
AK012111.1 | 9 | 978 | BAB28040.2 | |
AK020457.1 | ||||
BC010199.1 | 9 | 965 | ||
BC034174.1 | 9 | 988 | ||
BC039939.1 | 9 | 988 | AAH39939.1 | |
BC051052.1 | 9 | 989 | AAH51052.1 | |
BC060130.1 | 9 | 975 | AAH60130.1 | |
BC079652.1 | 9 | 961 | AAH79652.1 |
More Information
Set Membership: |
Intragenic Exonic |