Assay Details
Target Gene Details
Entrez Gene ID: | 22295 |
Gene Name: | cadherin 23 (otocadherin) |
Gene Aliases: |
4930542A03Rik, USH1D, ahl, ahl1, bob, bus, mdfw, nmf112, nmf181, nmf252, sals, v |
Location: |
Chr.10:60302414-60696515 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 69 - Exon 70 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cdh23 | NM_001252635.1 | NP_001239564.1 | ||
NM_023370.3 | NP_075859.2 | |||
NR_045556.1 | ||||
AF308939.1 | AAG52817.1 | |||
AJ888000.1 | 66 | 9947 | CAI59609.1 | |
AJ888001.1 | CAI59610.1 | |||
AJ888002.1 | CAI59611.1 | |||
AK039126.1 | ||||
AY026062.1 | AAK07670.1 | |||
AY563159.1 | AAT72159.1 | |||
AY563160.1 | AAT72160.1 | |||
AY563163.1 | AAT72163.1 | |||
AY563164.1 | AAT72164.1 |
More Information
Set Membership: |
Intragenic Non-exonic |