Assay Details
Target Gene Details
Entrez Gene ID: | 67500 |
Gene Name: | cell division cycle and apoptosis regulator 1 |
Gene Aliases: |
2610511G16Rik, 9430036H15Rik, Carp1 |
Location: |
Chr.10:62743928-62792368 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccar1 | NM_026201.3 | 24 | 3475 | NP_080477.1 |
AF465615.1 | 24 | 3366 | AAO17318.1 | |
AK077151.1 | 8 | 1176 | BAC36646.1 | |
AK078424.1 | 10 | 1452 | BAC37267.1 | |
BC079652.1 | 24 | 3449 | AAH79652.1 |
More Information
Set Membership: |
Intragenic Exonic |