Assay Details
Target Gene Details
Entrez Gene ID: | 66043 |
Gene Name: | ATP synthase, H+ transporting, mitochondrial F1 complex, delta subunit |
Gene Aliases: |
0610008F14Rik, 1500000I11Rik, AA960090, AI876556, AU020773, C85518 |
Location: |
Chr.10:80142280-80145818 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Atp5d | NM_025313.2 | NP_079589.2 | ||
AK002330.1 | BAB22016.1 | |||
AK005082.1 | ||||
AK011374.1 | BAB27577.1 | |||
AK018011.1 | BAB31035.1 | |||
AK076064.1 | ||||
AK151151.1 | BAE30157.1 | |||
AK155285.1 | BAE33165.1 | |||
AK165932.1 | BAE38468.1 | |||
AK186963.1 | ||||
BC008273.1 | AAH08273.1 | |||
BY451544.1 | ||||
CT010177.1 | CAJ18385.1 |
More Information
Set Membership: |
Intragenic Non-exonic |