Assay Details
Target Gene Details
Entrez Gene ID: | 28169 |
Gene Name: | 1-acylglycerol-3-phosphate O-acyltransferase 3 |
Gene Aliases: |
AW061257, AW493985, D10Jhu12e, lpaat3 |
Location: |
Chr.10:78269174-78352484 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Agpat3 | NM_053014.3 | NP_443747.2 | ||
AB377215.1 | ||||
AK008965.1 | ||||
AK015906.1 | BAB30025.2 | |||
AK030607.1 | BAC27043.1 | |||
AK075715.1 | BAC35905.1 | |||
AK076414.1 | BAC36329.1 | |||
AK159110.1 | BAE34826.1 | |||
AY167588.1 | AAN75574.1 | |||
BC052382.1 | AAH52382.1 | |||
BC058519.1 | AAH58519.1 |
More Information
Set Membership: |
Intragenic Non-exonic |