Assay Details
Target Gene Details
Entrez Gene ID: | 13115 |
Gene Name: | cytochrome P450, family 27, subfamily b, polypeptide 1 |
Gene Aliases: |
Cp2b, Cyp1, Cyp27b, Cyp40, P450c1, Pddr, Vdd1, Vddr, VddrI, Vdr |
Location: |
Chr.10:127048246-127053422 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cyp27b1 | NM_010009.2 | 8 | 1310 | NP_034139.2 |
AB006034.1 | 8 | 1304 | BAA22434.1 | |
BC080697.1 | 8 | 1345 | AAH80697.1 |
More Information
Set Membership: |
Intragenic Exonic |