Assay Details
Target Gene Details
Entrez Gene ID: | 22295 |
Gene Name: | cadherin 23 (otocadherin) |
Gene Aliases: |
4930542A03Rik, USH1D, ahl, ahl1, bob, bus, mdfw, nmf112, nmf181, nmf252, sals, v |
Location: |
Chr.10:60302414-60696515 on Build GRCm38 |
Assay Gene Location: | Within Exon 57 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cdh23 | NM_001252635.1 | 56 | 8413 | NP_001239564.1 |
NM_023370.3 | 57 | 8419 | NP_075859.2 | |
NR_045556.1 | 56 | 8413 | ||
AF308939.1 | 55 | 8012 | AAG52817.1 | |
AJ888000.1 | 54 | 8006 | CAI59609.1 | |
AJ888001.1 | 54 | 8006 | CAI59610.1 | |
AJ888002.1 | 54 | 8006 | CAI59611.1 | |
AK039126.1 | 12 | 1754 | ||
AY026062.1 | 55 | 8021 | AAK07670.1 | |
AY563163.1 | 9 | 1634 | AAT72163.1 | |
AY563164.1 | 9 | 1634 | AAT72164.1 |
More Information
Set Membership: |
Intragenic Exonic |