Assay Details
Target Gene Details
Entrez Gene ID: | 110279 |
Gene Name: | breakpoint cluster region |
Gene Aliases: |
5133400C09Rik, AI561783, AI853148, mKIAA3017 |
Location: |
Chr.10:75060557-75184923 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bcr | NM_001081412.2 | 13 | 2785 | NP_001074881.1 |
AK129482.1 | 10 | 975 | BAC98292.1 | |
AK183908.1 | 3 | 156 | ||
BC048842.1 | AAH48842.1 | |||
BC060270.1 | 13 | 2023 | AAH60270.1 | |
BC131682.1 | 14 | 2190 |
More Information
Set Membership: |
Intragenic Exonic |