Assay Details
Target Gene Details
Entrez Gene ID: | 72567 |
Gene Name: | BCL2-associated transcription factor 1 |
Gene Aliases: |
2610102K23Rik, 2700025J07Rik, 2810454G14Rik, 5730534O06Rik, AI450190, AW556225, Btf, mKIAA0164 |
Location: |
Chr.10:20311461-20342503 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bclaf1 | NM_001025392.1 | 6 | 2017 | NP_001020563.1 |
NM_001025393.1 | 6 | 2017 | NP_001020564.1 | |
NM_153787.2 | 6 | 2011 | NP_722482.1 | |
AK012290.1 | 1 | 803 | ||
AK079110.1 | 2 | 267 | BAC37542.1 | |
AK088741.1 | 6 | 1952 | BAC40542.1 | |
AK129071.1 | 3 | 1301 | BAC97881.1 | |
AK141834.1 | 6 | 1964 | BAE24851.1 | |
AK149724.1 | 6 | 1970 | BAE29048.1 | |
AK150017.1 | 6 | 1970 | BAE29242.1 | |
AK161692.1 | 6 | 2015 | BAE36535.1 | |
BC034300.1 | 6 | 1943 | AAH34300.1 |
More Information
Set Membership: |
Intragenic Exonic |