Assay Details
Target Gene Details
Entrez Gene ID: | 16467 |
Gene Name: | ataxia, cerebellar, Cayman type homolog (human) |
Gene Aliases: |
3322401A10Rik, BB077577, BNIP-H, CLAC, hes, ji |
Location: |
Chr.10:81204513-81230779 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Atcay | NM_178662.3 | NP_848777.1 | ||
AK028298.1 | BAC25867.1 | |||
AK038483.1 | BAC30014.1 | |||
AK038990.1 | BAC30195.1 | |||
AK039411.1 | BAC30342.1 | |||
AK086723.1 | ||||
AK162959.1 | BAE37136.1 | |||
AY349150.1 | AAQ90064.1 | |||
BC048903.1 | AAH48903.1 |