Assay Details
Target Gene Details
Entrez Gene ID: | 57267 |
Gene Name: | amyloid beta (A4) precursor protein-binding, family A, member 3 |
Gene Aliases: |
AW208791, Mint-3, X11gamma, lin-10, mint3 |
Location: |
Chr.10:81266965-81273247 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Apba3 | NM_018758.2 | NP_061228.1 | ||
AF070975.1 | AAC78837.1 | |||
AK045965.1 | BAC32549.1 | |||
AK169462.1 | BAE41187.1 | |||
AK170182.1 | BAE41620.1 | |||
BC005423.1 | AAH05423.1 | |||
BC009666.1 | AAH09666.1 | |||
BQ031518.1 |
More Information
Set Membership: |
Intragenic Non-exonic |