Assay Details
Target Gene Details
Entrez Gene ID: | 72567 |
Gene Name: | BCL2-associated transcription factor 1 |
Gene Aliases: |
2610102K23Rik, 2700025J07Rik, 2810454G14Rik, 5730534O06Rik, AI450190, AW556225, Btf, mKIAA0164 |
Location: |
Chr.10:20311461-20342503 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bclaf1 | NM_001025392.1 | NP_001020563.1 | ||
NM_001025393.1 | NP_001020564.1 | |||
NM_153787.2 | NP_722482.1 | |||
AK011802.1 | BAB27851.1 | |||
AK088741.1 | BAC40542.1 | |||
AK141834.1 | BAE24851.1 | |||
AK149724.1 | BAE29048.1 | |||
AK150017.1 | BAE29242.1 | |||
AK161692.1 | BAE36535.1 | |||
AK162896.1 | BAE37104.1 | |||
BC028963.1 | ||||
BC034300.1 | AAH34300.1 | |||
BY071148.1 |
More Information
Set Membership: |
Intragenic Non-exonic |