Assay Details
Target Gene Details
Entrez Gene ID: | 103220 |
Gene Name: | tetratricopeptide repeat domain 41 |
Gene Aliases: |
AI449705, AU041509, Gnn |
Location: |
Chr.10:86705632-86776844 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ttc41 | NM_001003910.2 | NP_001003910.2 | ||
NM_001003939.2 | NP_001003939.2 | |||
AK133195.1 | ||||
AY651019.1 | AAT81231.1 | |||
BC030307.1 | AAH30307.1 |