Assay Details
Target Gene Details
Entrez Gene ID: | 11702 |
Gene Name: | S-adenosylmethionine decarboxylase 1 |
Gene Aliases: |
1, AdoMetDC, Amd-1, SAMDC, SAMDC 1, adoMetDC1 |
Location: |
Chr.10:40287458-40302188 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Amd1 | NM_009665.5 | NP_033795.1 | ||
AK146840.1 | BAE27473.1 | |||
AK166187.1 | BAE38617.1 | |||
AK168165.1 | BAE40126.1 | |||
AK171238.1 | ||||
BC011110.1 | AAH11110.1 | |||
BC071220.1 | AAH71220.1 | |||
BC080791.1 | AAH80791.1 | |||
BC092072.1 | AAH92072.1 | |||
BC138696.1 | ||||
BC138697.1 | ||||
CT010192.1 | CAJ18400.1 | |||
D12780.1 | BAA02243.1 | |||
Z14986.1 | CAA78710.1 |
More Information
Set Membership: |
Intragenic Non-exonic |