Assay Details
Target Gene Details
Entrez Gene ID: | 216188 |
Gene Name: | aldehyde dehydrogenase 1 family, member L2 |
Gene Aliases: |
D330038I09Rik, mtFDH |
Location: |
Chr.10:83487283-83534146 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Aldh1l2 | NM_153543.2 | 9 | 1216 | NP_705771.2 |
AK052361.1 | 9 | 1107 | ||
AK086413.1 | 9 | 1160 | ||
AK141954.1 | 9 | 1178 | ||
AK188745.1 | 2 | 105 | ||
AK208193.1 | 2 | 105 | ||
BC034531.1 | 9 | 1138 | AAH34531.1 |
More Information
Set Membership: |
Intragenic Exonic |