Assay Details
Target Gene Details
Entrez Gene ID: | 216188 |
Gene Name: | aldehyde dehydrogenase 1 family, member L2 |
Gene Aliases: |
D330038I09Rik, mtFDH |
Location: |
Chr.10:83487283-83534146 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Aldh1l2 | NM_153543.2 | 3 | 443 | NP_705771.2 |
AK052361.1 | 3 | 334 | ||
AK086413.1 | 3 | 385 | ||
AK141954.1 | 3 | 405 | ||
AK192503.1 | ||||
AK201197.1 | ||||
AK203320.1 | ||||
BC034531.1 | 3 | 365 | AAH34531.1 | |
CJ130150.1 |
More Information
Set Membership: |
Intragenic Exonic |