Assay Details
Target Gene Details
Entrez Gene ID: | 11776 |
Gene Name: | adaptor-related protein complex 3, delta 1 subunit |
Gene Aliases: |
AA407035, Ap3d, Bolvr, mBLVR1, mh, mocha |
Location: |
Chr.10:80706956-80742282 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 22 - Exon 23 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ap3d1 | NM_007460.1 | NP_031486.1 | ||
AB004305.1 | BAA24578.1 | |||
AK187726.1 | ||||
BC048786.1 | AAH48786.1 | |||
BC053066.1 | AAH53066.1 |
More Information
Set Membership: |
Intragenic Non-exonic |