Assay Details
Target Gene Details
Entrez Gene ID: | 71712 |
Gene Name: | DNA-damage regulated autophagy modulator 1 |
Gene Aliases: |
1200002N14Rik, Dram |
Location: |
Chr.10:88322801-88364383 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dram1 | NM_027878.2 | NP_082154.2 | ||
AK004552.1 | BAB23366.2 | |||
AK151184.1 | ||||
BC021433.1 | AAH21433.1 |