Assay Details
Target Gene Details
Entrez Gene ID: | 116848 |
Gene Name: | bromodomain adjacent to zinc finger domain, 2A |
Gene Aliases: |
AA415431, C030005G16Rik, C78388, Tip5, Walp3, mKIAA0314 |
Location: |
Chr.10:128091056-128129304 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Baz2a | NM_054078.2 | 22 | 4252 | NP_473419.2 |
AJ309544.2 | 20 | 3986 | CAC69992.1 | |
AK122243.1 | 5 | 1098 | BAC65525.1 | |
AK135732.1 | 2 | 262 | BAE22634.1 | |
AK140558.1 | 1 | 69 | BAE24422.1 |
More Information
Set Membership: |
Intragenic Exonic |